Anti-GB3 Mouse Monoclonal Antibody [clone: BGR23]

Supplier: TCI America
A2506-1VIAL
TCA2506-VIALEA 705.1 USD
TCA2506-VIAL
Anti-GB3 Mouse Monoclonal Antibody [clone: BGR23]
Antibodies
Purified mouse monoclonal antibody/IgG2b, κ-chain

Type: Primary
Antigen: GB3
Clonality: Monoclonal
Clone: BGR23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR