Anti-GM1 Mouse Monoclonal Antibody [clone: GMB16]

Supplier: TCI America
A2505-1VIAL
TCA2505-VIALEA 631.9 USD
TCA2505-VIAL
Anti-GM1 Mouse Monoclonal Antibody [clone: GMB16]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: GM1
Clonality: Monoclonal
Clone: GMB16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR