4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥98.0% (by HPLC) for biochemical research

Supplier: TCI America

A2412-5MG
TCA2412-5MGEA 84.77 USD
TCA2412-5MG
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥98.0% (by HPLC) for biochemical research
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

[for Biochemical Research]
CAS Number: 28718-90-3
MDL Number: MFCD00012681
Molecular Formula: C16H15N5
Molecular Weight: 350.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Bioscience, Stains & Dyes, Fluorescent Stains
Other Product Names / Synonyms: 4',6-Diamidino-2-phenylindole Dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride

Formula: C₁₆H₁₅N₅·2HCl
MDL Number: MFCD00012681
CAS Number: 28718-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dd970dbedf5b446c85eabd71be8cf76412dbda7&vid=1a50cbeef1fdd1280d49ae3facb5454540e33233&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR