2-Azidoethyl-β-D-glucopyranoside ≥98.0%
Supplier: TCI America
Synonyms:
β-D-Glc-Et-N3
A2267-1G
TCA2267-1GEA
1106.8
USD
TCA2267-1G
2-Azidoethyl-β-D-glucopyranoside ≥98.0%
2-Azidoethyl-beta-D-glucopyranoside
CAS Number: 165331-08-8
Molecular Formula: C8H15N3O6
Molecular Weight: 249.22
Purity/Analysis Method:
Formula:
C₈H₁₅N₃O₆ |
CAS Number:
165331-08-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...