2-Amino-6-chloropyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2263-5G
TCA2263-5GEA
73
USD
TCA2263-5G
2-Amino-6-chloropyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-chloropyridine
CAS Number: 45644-21-1
MDL Number: MFCD00234068
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method:
Formula:
C₅H₅ClN₂ Melting Pt: 72 °C |
MDL Number:
MFCD00234068 CAS Number: 45644-21-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...