6-Aminocaproic acid ≥98.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
Synonyms:
6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid
A2255-25G
A2255-5G
TCA2255-25GEA
76.19
USD
TCA2255-25G
TCA2255-5G
6-Aminocaproic acid ≥98.0% (by titrimetric analysis) for biochemical research
6-Aminocaproic acid
[for Biochemical Research]
CAS Number: 60-32-2
MDL Number: MFCD00008238
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 204
Storage Temperature: 0-10°C
Formula:
C₆H₁₃NO₂ Melting Pt: 204 °C |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...