4-Aminoantipyrine ≥98.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

A2254-1G A2254-5G
TCA2254-1GEA 26 USD
TCA2254-1G TCA2254-5G
4-Aminoantipyrine ≥98.0% (by titrimetric analysis) for biochemical research
4-Aminoantipyrine

[for Biochemical Research]
CAS Number: 83-07-8
MDL Number: MFCD00003145
Molecular Formula: C11H13N3O
Molecular Weight: 203.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 109


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 4-Aminophenazone, 4-AA

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 109 °C
MDL Number: MFCD00003145
CAS Number: 83-07-8
Merck Index: 13,00596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c8d644bd64bc916afd15d4ee016e47da6718bef&vid=103d4393579d3958fad24c16e6b6837715df1aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR