4-Aminopyrimidine ≥98.0%
Supplier: TCI America
A2251-1G
TCA2251-1GEA
138
USD
TCA2251-1G
4-Aminopyrimidine ≥98.0%
4-Aminopyrimidine
CAS Number: 591-54-8
MDL Number: MFCD00006112
Molecular Formula: C4H5N3
Molecular Weight: 95.11
Purity/Analysis Method:
Formula:
C₄H₅N₃ MW: 95.1 Melting Pt: 152 °C |
MDL Number:
MFCD00006112 CAS Number: 591-54-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...