Allylboronic acid pinacol ester ≥96.0% stabilised
Supplier: TCI America
Synonyms:
2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane
A2157-1G
A2157-5G
TCA2157-1GEA
42
USD
TCA2157-1G
TCA2157-5G
Allylboronic acid pinacol ester ≥96.0% stabilised
Allylboronic acid pinacol ester
(stabilized with Phenothiazine)
CAS Number: 72824-04-5
MDL Number: MFCD00013347
Molecular Formula: C9H17BO2
Molecular Weight: 168.04
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Allylboronic Acid Pinacol Ester
Formula:
C₉H₁₇BO₂ MW: 168.05 g/mol Boiling Pt: 35 °C Density: 0.89 (20 °C) Flash Pt: 46 °C |
MDL Number:
MFCD00013347 CAS Number: 72824-04-5 UN: 3272 ADR: 3 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...