Antazoline hydrochloride ≥99.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 2508-72-7
MDL Number: MFCD00058145
Molecular Formula: C17H19N3
Molecular Weight: 301.82
Purity/Analysis Method:
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: 2-(N-Benzylanilinomethyl)-2-imidazoline Hydrochloride
Formula:
C₁₇H₁₉N₃ Melting Pt: 238 °C |
MDL Number:
MFCD00058145 CAS Number: 2508-72-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...