Naphthalene black 10B (naphthol blue black, acid black 1) ≥97.0% (by HPLC)
Supplier: TCI America
CAS Number: 1064-48-8
MDL Number: MFCD00004017
Molecular Formula: C22H16N6O9S2
Molecular Weight: 616.49
Purity/Analysis Method:
Product Categories: Bioscience, Stains & Dyes, Analytical Science, Reagents for Staining and Others (Reagents for Electrophoresis)
Other Product Names / Synonyms: Amido Black 10B, Aminoschwarz 10B, Buffalo Black NBR, Naphthol Blue Black
Formula:
C₂₂H₁₄N₆Na₂O₉S₂ |
MDL Number:
MFCD00004017 CAS Number: 1064-48-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...