N4-Acetylcytosine ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Oxo-1,2-dihydropyrimidin-4-yl)acetamide

A2089-25G A2089-5G
TCA2089-25GEA 91 USD
TCA2089-25G TCA2089-5G
N4-Acetylcytosine ≥98.0%
N4-Acetylcytosine

CAS Number: 14631-20-0 MDL Number: MFCD00134466 Molecular Formula: C6H7N3O2 Molecular Weight: 153.14 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 325

Formula: C₆H₇N₃O₂
Melting Pt: 325 °C
MDL Number: MFCD00134466
CAS Number: 14631-20-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fad22d9b745d3c2c543791616391c500adda4f6c&vid=fad22d9b745d3c2c543791616391c500adda4f6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR