7-Aminodesacetoxycephalosporanic acid ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 22252-43-3
MDL Number: MFCD00151456
Molecular Formula: C8H10N2O3S
Molecular Weight: 214.24
Purity/Analysis Method:
Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: 7-ADCA
Formula:
C₈H₁₀N₂O₃S Melting Pt: 242 °C |
MDL Number:
MFCD00151456 CAS Number: 22252-43-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...