Ammonium chloride ≥99.5%
Supplier: TCI America
A2037-300G
TCA2037-300GEA
34.06
USD
TCA2037-300G
Ammonium chloride ≥99.5%
Ammonium chloride
CAS Number: 12125-02-9
MDL Number: MFCD00011420
Molecular Formula: H4ClN
Molecular Weight: 53.49
Purity/Analysis Method:
Formula:
NH₄Cl Boiling Pt: 520 °C Melting Pt: 338 °C |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...