Acetonitrile-[D₃] (99.8% D)

Supplier: TCI America

A2018-10ML
TCA2018-10MLEA 97 USD
TCA2018-10ML
Acetonitrile-[D₃] (99.8% D)
Acetonitrile-[D3]

CAS Number: 2206-26-0 MDL Number: MFCD00001881 Molecular Formula: C2D3N Molecular Weight: 44.07 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 81 Flash Point (°C): 6 Specific Gravity (20/20): 0.84


Product Categories: Analytical Science, Deuterated Compounds for NMR Other Product Names / Synonyms: Trideuteroacetonitrile, Methyl-d3 Cyanide

Formula: D₃CCN
Boiling Pt: 81 °C
Density: 0.84 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR