(R)-tert-Butyl-2-aminopropanoate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
CAS Number: 59531-86-1
MDL Number: MFCD00077087
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: tert-Butyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride, H-D-Ala-OtBu.HCl
Formula:
C₇H₁₅NO₂·HCl Melting Pt: 208 °C |
MDL Number:
MFCD00077087 CAS Number: 59531-86-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...