8-Acetyl-7-hydroxy-4-methylcoumarin ≥98.0%
Supplier: TCI America
CAS Number: 2555-29-5
Molecular Formula: C12H10O4
Molecular Weight: 218.21
Purity/Analysis Method:
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 8-Acetyl-4-methylumbelliferone
Formula:
C₁₂H₁₀O₄ Melting Pt: 170 °C |
CAS Number:
2555-29-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...