2-Anthraceneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
A1843-1G
A1843-5G
TCA1843-1GEA
154
USD
TCA1843-1G
TCA1843-5G
2-Anthraceneboronic acid (contains varying amounts of Anhydride)
2-Anthraceneboronic acid
CAS Number: 141981-64-8
MDL Number: MFCD07784001
Molecular Formula: C14H11BO2
Molecular Weight: 222.05
Form: Crystal
Color: Very Pale Yellow
Formula:
C₁₄H₁₁BO₂ |
MDL Number:
MFCD07784001 CAS Number: 141981-64-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...