Aluminium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1831-100G A1831-500G
TCA1831-100GEA 50.26 USD
TCA1831-100G TCA1831-500G
Aluminium chloride ≥98.0% (by titrimetric analysis)
Aluminium chloride

CAS Number: 7446-70-0 Molecular Formula: AlCl3 Molecular Weight: 133.33 Purity/Analysis Method: 98.0% (T) Form: Crystal

Formula: AlCl₃
MDL Number: MFCD00003422
CAS Number: 7446-70-0
UN: 1726
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd769378e55bda6b7937d31092d21168f8d3ee56&vid=bd769378e55bda6b7937d31092d21168f8d3ee56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR