p-Anisaldehyde in ethanol (contains Acetic Acid, H2SO4)

Supplier: TCI America

Synonyms: 4-Methoxybenzaldehyde

A1674-500ML
TCA1674-500MLEA 124.24 USD
TCA1674-500ML
p-Anisaldehyde in ethanol (contains Acetic Acid, H2SO4)
p-Anisaldehyde

[for TLC Stain]
CAS Number: 123-11-5
MDL Number: MFCD00003385
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Form: Clear Liquid
Color: Colorless

Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00003385
CAS Number: 123-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9785f5e5a82d119c6bd327473326b3f8da4ed64a&vid=64d2a02464457d0d82d716682b8899a53fc1ce20&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR