Acetaldehyde ∼2% in DMF

Supplier: TCI America

Synonyms: Ethanal

A1640-10ML
TCA1640-10MLEA 69.15 USD
TCA1640-10ML
Acetaldehyde ∼2% in DMF
Acetaldehyde

[for Detection of Primary and Secondary Amines]
CAS Number: 75-07-0
MDL Number: MFCD00006991
Molecular Formula: C2H4O
Molecular Weight: 44.05
Form: Clear Liquid
Color: Colorless

Formula: CH₃CHO
MW: 44.05 g/mol
MDL Number: MFCD00006991
CAS Number: 75-07-0
UN: 1089
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5b07aaac7cb312f7b9035077e42772009342904&vid=b4c7c96cdc0b0eedea5780f399f7aec84da0f876&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR