Ponceau S ≥85.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
CAS Number: 6226-79-5
MDL Number: MFCD00003892
Molecular Formula: C22H16N4O13S4
Molecular Weight: 760.55
Purity/Analysis Method:
Other Product Names / Synonyms: Ponceau S
Formula:
C₂₂H₁₂N₄Na₄O₁₃S₄ MW: 760.56 g/mol |
MDL Number:
MFCD00003892 CAS Number: 6226-79-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...