3-Methylanthranil acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 4389-45-1
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Amino-m-toluic Acid, 3-Methylanthranilic Acid
Formula:
C₈H₉NO₂ Melting Pt: 175 °C |
MDL Number:
MFCD00007745 CAS Number: 4389-45-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...