(S)-Methyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 2491-20-5
MDL Number: MFCD00063663
Molecular Formula: C4H9NO2
Molecular Weight: 139.58
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride, H-Ala-OMe.HCl
Formula:
C₄H₉NO₂·HCl Melting Pt: 108 °C |
MDL Number:
MFCD00063663 CAS Number: 2491-20-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...