L-α-Aminoadipic acid ≥98.0%
Supplier: TCI America
Synonyms:
L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid
A1457-1G
A1457-5G
TCA1457-001GEA
151.7
USD
TCA1457-001G
TCA1457-005G
L-α-Aminoadipic acid ≥98.0%
L-α-Aminoadipic acid
CAS Number: 1118-90-7
MDL Number: MFCD00002636
Molecular Formula: C6H11NO4
Molecular Weight: 161.16
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Aminohexanedioic Acid
Formula:
C₆H₁₁NO₄ MW: 161.16 g/mol |
MDL Number:
MFCD00002636 CAS Number: 1118-90-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...