Atenolol ≥98.0%
Supplier: TCI America
Synonyms:
2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide
A1414-10G
TCA1414-010GEA
224.21
USD
TCA1414-010G
Atenolol ≥98.0%
Atenolol
CAS Number: 29122-68-7
MDL Number: MFCD00057645
Molecular Formula: C14H22N2O3
Molecular Weight: 266.34
Purity/Analysis Method:
Product Categories: Bioscience, Antihypertensive Ingredients, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide
Formula:
C₁₄H₂₂N₂O₃ MW: 266.34 g/mol Melting Pt: 154 °C |
MDL Number:
MFCD00057645 CAS Number: 29122-68-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...