N-Boc-ethylenediamine ≥97.0% (by GC)

Supplier: TCI America

A1371-1G A1371-25G A1371-5G
TCA1371-1GEA 84.63 USD
TCA1371-1G TCA1371-25G TCA1371-5G
N-Boc-ethylenediamine ≥97.0% (by GC)
N-Boc-ethylenediamine

CAS Number: 57260-73-8 MDL Number: MFCD00191871 Molecular Formula: C7H16N2O2 Molecular Weight: 160.22 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 105 Flash Point (°C): 110 Specific Gravity (20/20): 1.01


Product Categories: Bioscience, Chemical Building Blocks for Polyamines, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method) Other Product Names / Synonyms: N-(2-Aminoethyl)carbamic Acid tert-Butyl Ester, N-Boc-1,2-diaminoethane, N-Boc-ethylenediamine, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl N-(2-Aminoethyl)carbamate

Formula: C₇H₁₆N₂O₂
Boiling Pt: 105 °C
Density: 1.01 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00191871
CAS Number: 57260-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7a7693aacc9255a354f2769be7f2407462d8bf5&vid=f7a7693aacc9255a354f2769be7f2407462d8bf5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR