2-Acetamido-6-hydroxypurine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 19962-37-9
MDL Number: MFCD00078201
Molecular Formula: C7H7N5O2
Molecular Weight: 193.17
Purity/Analysis Method:
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: N-Acetylguanine
Formula:
C₇H₇N₅O₂ |
MDL Number:
MFCD00078201 CAS Number: 19962-37-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...