1-Anthracenecarboxylic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Anthracenecarboxylic Acid

A1220-1G
TCA1220-001GEA 307.6 USD
TCA1220-001G
1-Anthracenecarboxylic acid ≥99.0% (by titrimetric analysis)
1-Anthracenecarboxylic acid

CAS Number: 607-42-1 MDL Number: MFCD00142583 Molecular Formula: C15H10O2 Molecular Weight: 222.24 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 252

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Melting Pt: 252 °C
MDL Number: MFCD00142583
CAS Number: 607-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82708abd302c728a5ff810f9e2d2ce20e521c414&vid=82708abd302c728a5ff810f9e2d2ce20e521c414&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR