3-Aminobenzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1203-25G A1203-5G
TCA1203-25GEA 181 USD
TCA1203-25G TCA1203-5G
3-Aminobenzonitrile ≥98.0% (by GC, titration analysis)
3-Aminobenzonitrile

CAS Number: 2237-30-1 MDL Number: MFCD00007756 Molecular Formula: C7H6N2 Molecular Weight: 118.14 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 290 Melting point (°C): 53 Flash Point (°C): 110


Other Product Names / Synonyms: 3-Cyanoaniline

Formula: C₇H₆N₂
Boiling Pt: 290 °C
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD00007756
CAS Number: 2237-30-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0ab98d31a38b8fd321df79cd89cdbbe1276a29f&vid=53a36d861d523f6f29c25eeaab939fb4f2312f67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR