Diethyl aminomalonate hydrochloride

Supplier: TCI America

Synonyms: Diethyl 2-aminomalonate hydrochloride

A1016-25G A1016-500G A1016-100G
TCA1016-025GEA 17 USD
TCA1016-025G TCA1016-500G TCA1016-100G
Diethyl aminomalonate hydrochloride
Diethyl aminomalonate hydrochloride

CAS Number: 13433-00-6 MDL Number: MFCD00012510 Molecular Formula: C7H13NO4 Molecular Weight: 211.64 Form: Crystal Color: White


Other Product Names / Synonyms: Aminomalonic Acid Diethyl Ester Hydrochloride

Formula: C₇H₁₄ClNO₄
MW: 211.65 g/mol
MDL Number: MFCD00012510
CAS Number: 13433-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebb3e9d7e42de546bf3e61c722fa35eecf3b2af&vid=bebb3e9d7e42de546bf3e61c722fa35eecf3b2af&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR