5-Aminoorotic acid ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
5-Aminouracil-6-carboxylic acid, 5-Aminoorotic Acid, 5-Aminouracil-6-carboxylic Acid, 5-Amino-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
A0947-25G
TCA0947-025GEA
131.33
USD
TCA0947-025G
5-Aminoorotic acid ≥97.0% (by total nitrogen and titration analysis)
5-Aminoorotic acid
CAS Number: 7164-43-4
MDL Number: MFCD00010563
Molecular Formula: C5H5N3O4
Molecular Weight: 171.11
Purity/Analysis Method:
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 5-Aminouracil-6-carboxylic Acid
Formula:
C₅H₅N₃O₄ MW: 171.11 g/mol |
MDL Number:
MFCD00010563 CAS Number: 7164-43-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...