Allura red AC

Supplier: TCI America

A0943-25G A0943-5G A0943-100G
TCA0943-25GEA 69.56 USD
TCA0943-25G TCA0943-005G TCA0943-100G
Allura red AC
Allura red AC

Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: FD and C Red No.40

Formula: C₁₈H₁₄N₂O₈S₂·2Na
MDL Number: MFCD00059526
CAS Number: 25956-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff483414c19e762e17c6929263e7848024ca6c85&vid=ff483414c19e762e17c6929263e7848024ca6c85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Lambda max. 497.0 to 501.0 nm (0.02mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 400 (0.02mol/L AcONH4 sol., 497.0 to 501.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR