4-Nitro-2-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America
Warning

A0805-25G A0805-1G A0805-5G
TCA0805-25GEA 51 USD
TCA0805-25G TCA0805-1G TCA0805-5G
4-Nitro-2-(trifluoromethyl)aniline ≥98.0%
4-Nitro-2-(trifluoromethyl)aniline

CAS Number: 121-01-7 MDL Number: MFCD00007365 Molecular Formula: C7H5F3N2O2 Molecular Weight: 206.12 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 91


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: 4-Nitro-2-trifluoromethylaniline

Formula: C₇H₅F₃N₂O₂
Melting Pt: 91 °C
Density: 1.754 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a5e0b90fd1cd71993556ba07b7a7b097bc1967f&vid=0a5e0b90fd1cd71993556ba07b7a7b097bc1967f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR