2,6-Di-O-palmitoyl-L-ascorbic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
L-Ascorbyl 2,6-Dipalmitate
A0757-25G
TCA0757-025GEA
102.5
USD
TCA0757-025G
2,6-Di-O-palmitoyl-L-ascorbic acid ≥97.0% (by titrimetric analysis)
2,6-Di-O-palmitoyl-L-ascorbic acid
CAS Number: 4218-81-9
MDL Number: MFCD00059738
Molecular Formula: C38H68O8
Molecular Weight: 652.95
Purity/Analysis Method:
Product Categories: Bioscience, Sugar Acids, Sugars (Others), Vitamin Derivatives
Other Product Names / Synonyms: L-Ascorbyl 2,6-Dipalmitate
Formula:
C₃₈H₆₈O₈ MW: 652.95 g/mol Melting Pt: 113 °C |
MDL Number:
MFCD00059738 CAS Number: 4218-81-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...