2-Adamantanol ≥98.0%
Supplier: TCI America
A0720-5G
A0720-25G
TCA0720-005GEA
43
USD
TCA0720-005G
TCA0720-025G
2-Adamantanol ≥98.0%
2-Adamantanol
CAS Number: 700-57-2
MDL Number: MFCD00074744
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Hydroxyadamantane
Formula:
C₁₀H₁₆O Melting Pt: 299…303 °C Density: 1.07 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00074744 CAS Number: 700-57-2 EINECS: 211-846-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...