2-Adamantylamine hydrochloride ≥99.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 10523-68-9
MDL Number: MFCD00074743
Molecular Formula: C10H17N
Molecular Weight: 187.71
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Aminoadamantane Hydrochloride
Formula:
C₁₀H₁₈ClN Melting Pt: 280 °C |
MDL Number:
MFCD00074743 CAS Number: 10523-68-9 EINECS: 234-074-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...