(±)-3-Amino-1,2-propanediol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A0628-25G
TCA0628-25GEA
59
USD
TCA0628-25G
(±)-3-Amino-1,2-propanediol ≥98.0% (by GC, titration analysis)
(±)-3-Amino-1,2-propanediol
CAS Number: 616-30-8
MDL Number: MFCD00008140
Molecular Formula: C3H9NO2
Molecular Weight: 91.11
Purity/Analysis Method:
Formula:
C₃H₉NO₂ Flash Pt: 155 °C |
MDL Number:
MFCD00008140 CAS Number: 616-30-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...