6-O-Stearoyl-L-ascorbic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
L-Ascorbyl 6-Stearate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl stearate
A0617-5G
TCA0617-5GEA
144
USD
TCA0617-5G
6-O-Stearoyl-L-ascorbic acid ≥95.0% (by titrimetric analysis)
6-O-Stearoyl-L-ascorbic acid
CAS Number: 10605-09-1
MDL Number: MFCD00059739
Molecular Formula: C24H42O7
Molecular Weight: 442.59
Purity/Analysis Method:
Product Categories: Bioscience, Sugar Acids, Sugars (Others), Vitamin Derivatives
Other Product Names / Synonyms: L-Ascorbyl 6-Stearate
Formula:
C₂₄H₄₂O₇ MW: 442.59 g/mol Melting Pt: 117 °C |
MDL Number:
MFCD00059739 CAS Number: 10605-09-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...