Acid red 1
Supplier: TCI America
CAS Number: 3734-67-6
MDL Number: MFCD00003954
Molecular Formula: C18H15N3O8S2
Molecular Weight: 509.41
Form: Powder
Color: Red
Other Product Names / Synonyms: Amidonaphthol Red, Azophloxine, Pontacyl Carmine, Red 2G
Formula:
C₁₈H₁₅N₃O₈S₂ |
MDL Number:
MFCD00003954 CAS Number: 3734-67-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...