L(+)-Aspartic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0546-25G A0546-500G
TCA0546-025GEA 29.29 USD
TCA0546-025G TCA0546-500G
L(+)-Aspartic acid ≥99.0% (by titrimetric analysis)
L(+)-Aspartic acid

CAS Number: 56-84-8 MDL Number: MFCD00002616 Molecular Formula: C4H7NO4 Molecular Weight: 133.10 Purity/Analysis Method: 99.0% (T) Form: Crystal


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids Other Product Names / Synonyms: (S)-Aminosuccinic Acid, L-Asparaginic Acid, H-Asp-OH

Formula: C₄H₇NO₄
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 251 °C
Density: 1.6613 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=803bff47aaf667a2bd3a97876712dd529387367f&vid=803bff47aaf667a2bd3a97876712dd529387367f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR