(±)-Aspartic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Aminobutanedioic acid, DL-Aspartic acid

A0544-25G A0544-500G
TCA0544-025GEA 22.83 USD
TCA0544-025G TCA0544-500G
(±)-Aspartic acid ≥98.0%
(±)-Aspartic acid

CAS Number: 617-45-8 MDL Number: MFCD00063083 Molecular Formula: C4H7NO4 Molecular Weight: 133.10 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 280


Product Categories: Bioscience, Proteinogenic Amino Acids Other Product Names / Synonyms: Aminosuccinic Acid, DL-Asparaginic Acid, H-DL-Asp-OH

Formula: C₄H₇NO₄
MW: 133.10 g/mol
Melting Pt: 280 °C
MDL Number: MFCD00063083
CAS Number: 617-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db43af7b2b1a1e16a9aa458b67204344fb535c0d&vid=115d75703194ca8611108689871c6d34dc527549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR