Arbutin ≥95.0%
Supplier: TCI America
Synonyms:
4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside
CAS Number: 497-76-7
MDL Number: MFCD00016915
Molecular Formula: C12H16O7
Molecular Weight: 272.25
Purity/Analysis Method:
Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Crude Medicine Ingredients for Research and Experimental Use, Cosmetic Ingredients for Research and Experimental Use, Others (Phytochemicals)
Other Product Names / Synonyms: Hydroquinone-beta-D-glucoside, 4-Hydroxyphenyl beta-D-Glucopyranoside
Formula:
C₁₂H₁₆O₇ MW: 272.25 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00016915 CAS Number: 497-76-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...