8-Anilino-1-naphthalenesulfonic acid ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 82-76-8
MDL Number: MFCD00003998
Molecular Formula: C16H13NO3S
Molecular Weight: 299.34
Purity/Analysis Method:
Product Categories: Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Phenyl Acid, N-Phenyl-1-naphthylamine-8-sulfonic Acid, Phenyl Peri Acid
Formula:
C₁₆H₁₃NO₃S Melting Pt: 217 °C |
MDL Number:
MFCD00003998 CAS Number: 82-76-8 EINECS: 201-438-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...