Aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Aminobenzene, Phenylamine

A0463-500G
TCA0463-500GEA 53.87 USD
TCA0463-500G
Aniline ≥98.0% (by GC, titration analysis)
Aniline

CAS Number: 62-53-3 MDL Number: MFCD00007629 Molecular Formula: C6H7N Molecular Weight: 93.13 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 184 Melting point (°C): -6 Flash Point (°C): 75 Specific Gravity (20/20): 1.02

Formula: C₆H₅NH₂
MW: 93.13 g/mol
Boiling Pt: 184 °C
Melting Pt: –6 °C
Density: 1.02 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00007629
CAS Number: 62-53-3
UN: 1547
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3bc10ff0288aeca7acf0874c1b2e6d47bfe3e32&vid=d3bc10ff0288aeca7acf0874c1b2e6d47bfe3e32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR