4-Aminopyridine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

A0414-500G A0414-25G A0414-100G
TCA0414-500GEA 296 USD
TCA0414-500G TCA0414-25G TCA0414-100G
4-Aminopyridine ≥99.0% (by GC, titration analysis)
4-Aminopyridine

CAS Number: 504-24-5 MDL Number: MFCD00006439 Molecular Formula: C5H6N2 Molecular Weight: 94.12 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Boiling point (°C): 287 Melting point (°C): 158 Flash Point (°C): 164

Formula: C₅H₆N₂
Boiling Pt: 287 °C
Melting Pt: 158 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS Number: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 13,03964
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9af5cbcacf6af70e3eb96565ed53ef0cd81fd83&vid=b9af5cbcacf6af70e3eb96565ed53ef0cd81fd83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR