4-Aminopyridine ≥99.0%
Supplier: TCI America
Synonyms:
Pyridin-4-amine, 4-Pyridylamine
A0414-500G
A0414-25G
A0414-100G
TCA0414-500GEA
296
USD
TCA0414-500G
TCA0414-25G
TCA0414-100G
4-Aminopyridine ≥99.0%
4-Aminopyridine
CAS Number: 504-24-5
MDL Number: MFCD00006439
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method:
Formula:
C₅H₆N₂ Boiling Pt: 287 °C Melting Pt: 158 °C Density: 1.25 g/cm³ (20 °C) Flash Pt: 164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 EINECS: 207-987-9 UN: 2671 ADR: 6.1,II Merck Index: 13,03964 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...