2-Aminopyridine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

A0411-500G A0411-25G A0411-100G
TCA0411-500GEA 66 USD
TCA0411-500G TCA0411-25G TCA0411-100G
2-Aminopyridine ≥99.0% (by GC, titration analysis)
2-Aminopyridine

CAS Number: 504-29-0 MDL Number: MFCD00006312 Molecular Formula: C5H6N2 Molecular Weight: 94.12 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Boiling point (°C): 105 Melting point (°C): 61 Flash Point (°C): 68 Storage Temperature: 0-10°C

Formula: C₅H₆N₂
Boiling Pt: 105 °C
Melting Pt: 61 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3822a3ec11cdee4f04df5e5a1de2399015efc8d3&vid=3822a3ec11cdee4f04df5e5a1de2399015efc8d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR