2-Amino-3-methylpyridine ≥98.0%

Supplier: TCI America

A0400-25G
TCA0400-500GEA 240.46 USD
TCA0400-500G TCA0400-25G
2-Amino-3-methylpyridine ≥98.0%
2-Amino-3-methylpyridine

CAS Number: 1603-40-3 MDL Number: MFCD00006320 Molecular Formula: C6H8N2 Molecular Weight: 108.14 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 221 Flash Point (°C): 118 Freezing point (°C): 31


Other Product Names / Synonyms: 2-Amino-3-picoline

Formula: C₆H₈N₂
Boiling Pt: 221 °C
Melting Pt: 29…32 °C
Density: 1.073 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02ac60fb48f013b0b66a2d9cd3fbb0e99bf5de31&vid=02ac60fb48f013b0b66a2d9cd3fbb0e99bf5de31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR