4-Aminophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 123-30-8
MDL Number: MFCD00007869
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method:
Formula:
H₂NC₆H₄OH Boiling Pt: 284 °C Melting Pt: 187 °C Density: 1.3 g/cm³ (20 °C) Flash Pt: 195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007869 CAS Number: 123-30-8 EINECS: 204-616-2 UN: 2512 ADR: 6.1,III Merck Index: 13,00461 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...